n-Hexane 95+% ACS
Glass bottle, 1 L
: Hexyl hydride, Hexanen, Hexane
- :110-54-3
- :H₃C(CH₂)₄CH₃
- :MFCD00009520
- :86.18 g/mol
- :69 °C (760 mmHg)
- :–95 °C
- :0.659 g/cm³
- :–22 °C
- :Ambient
- :1208
- :3,II
- Appearance:Clear liquid
- GC:≥95.0 % (n-Hexane)
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Sulfur compounds (as S):≤0.005 %
- Water soluble titr. acid:≤0.0003 meg/g
- Thiophene:Passes test
- Pack type:Glass bottle
- Pk:1 L



