Orders LinkContactUsLinkComponent
VWR®, Overalls, SPP
VWR®, Overalls, SPP
White, L
Catalog # 113-8312
Supplier:   VWR Collection
VWR®, Overalls, SPP
Catalog # 113-8312
Supplier:  VWR Collection
VWR®, Overalls, SPP
VWR®, Overalls, SPP
White, L
Price
106,00 €
Log in for availability
1 * 25 items
Product Details & Documents
Breathable overalls made of spunbonded polypropylene (SPP), a lightweight fabric that provides protection in non hazardous, dirty work environments.
  • With collar
  • Two-way nylon zip
  • Elasticated cuffs and ankles
Weight: 40 g/m².
Colour: White.
Specifications
  • Clothing Size:
    L
  • Colour:
    White
  • Seq:
    02
  • Cat. No.:
    113-8312
  • Model:
    With collar
  • Garment application:
    General Purpose|Industrial
  • Weight:
    40 g/m²