Orders LinkContactUsLinkComponent
Leather gloves, A271
Leather gloves, A271
XL, Light brown
Catalog # PRTWA271TARXL
Supplier:   Portwest
Leather gloves, A271
Catalog # PRTWA271TARXL
Supplier:  Portwest
Leather gloves, A271
Leather gloves, A271
XL, Light brown
Price
151,00 €
Log in for availability
1 * 12 PAIR
Product Details & Documents
Premium quality leather drivers glove with Insulatex™ lining for added warmth and comfort.
  • Made from cow grain leather
  • Provide mechanical and cold protection
Certifications: EN 388:2016 (3142X), EN 511 (22X)
Specifications
  • Glove size:
    XL
  • Colour:
    Light brown
  • Seq:
    02
  • Cat. No.:
    PRTWA271TARXL
  • Liner material:
    Insulatex™
  • Disposable / reusable:
    Reusable
  • Model:
    A271
  • EN norm:
    EN 388:2016 (3142X), EN 511 (22X)
  • Ambidextrous / hand-specific:
    Hand-specific