ContactUsLinkComponent
safety storage cabinet, tall cabinet with wing doors, fixed window, model line q-line, model class q-display-30, model q30.195.086.w dfw, with cylinder locking exterior: wxdxh (mm) 864 x 620 x 1947 interior: wxdxh (mm) 750 x 519 x 1626 body ral9010 g 1 * 1 items
Catalog # 135-2074
Supplier:  asecos
safety storage cabinet, tall cabinet with wing doors, fixed window, model line q-line, model class q-display-30, model q30.195.086.w dfw, with cylinder locking exterior: wxdxh (mm) 864 x 620 x 1947 interior: wxdxh (mm) 750 x 519 x 1626 body ral9010 g 1 * 1 items
Catalog # 135-2074
Supplier:  asecos