Reagent troughs
Disposable reagent trough for maximum reagent recovery, Tecan Pure, 25 ml, Grey
Catalog # TCAN30055743
Supplier: Tecan
Jump to:
Product Details & Documents
These disposable PP troughs are designed to optimise the performance and productivity of Tecan liquid handling platforms.
Specifications
- Capacity:25 ml
- Colour:Grey
- Description:Disposable reagent trough for maximum reagent recovery, Tecan Pure
- Seq:01