Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium hydroxide 1 N in aqueous solution USP Test Solution (TS)

Sodium hydroxide 1 N in aqueous solution USP Test Solution (TS)

Plastic bottle, 100 mL
Catalog # 85373.180
Corrosive
Supplier:   VWR Chemicals
Sodium hydroxide 1 N in aqueous solution USP Test Solution (TS)
Catalog # 85373.180
Supplier:  VWR Chemicals
CAS Number:  1310-73-2
Sodium hydroxide 1 N in aqueous solution USP Test Solution (TS)
Sodium hydroxide 1 N in aqueous solution USP Test Solution (TS)

Plastic bottle, 100 mL

CAS Number:  1310-73-2
Price
29,20 €
Log in for availability
1 * 100 mL
Product Details & Documents

Ready to use solutions.

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Ambient
  • EINECS:0
  • UN:1824
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL

Specifications