Silver nitrate 0.1 N in aqueous solution USP Test Solution (TS)
Glass bottle, 500 mL
Supplier: VWR Chemicals
Silver nitrate 0.1 N in aqueous solution USP Test Solution (TS)
Catalog # 85370.260
Supplier: VWR Chemicals
CAS Number: 7761-88-8
Jump to:
Product Details & Documents
Ready to use solution.
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Merck Index:12,08661
- Melting Pt:210 °C
- Storage Temperature:Ambient
- EINECS:0
- UN:3264
- ADR:8,III
- REACH:01-2119513705-43
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL



