Order Entry
Export
Orders LinkContactUsLinkComponent
VWR®, Density standards referenced at 20 °C

VWR®, Density standards referenced at 20 °C

Density standard, 0,7374 g/ml, 20 °C, 100 mL
Catalog # 85174.180
Supplier:   VWR Chemicals
VWR®, Density standards referenced at 20 °C
Catalog # 85174.180
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Density standards referenced at 20 °C
VWR®, Density standards referenced at 20 °C

Density standard, 0,7374 g/ml, 20 °C, 100 mL

CAS Number:  
Price
107,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.

  • High accuracy across an extensive density range (0,6619 to 3,1096 g/ml) at 20 °C
  • No toxic heavy metals used in any formulation
  • Can be used with any brand or type of density measuring instrument
  • Supplied in a high quality, tamper-proof, amber glass bottle

These standards are prepared gravimetrically (solute and solvent) using OIML traceable weights. The density values of the standards are established using fully calibrated reference pycnometers.

Certifications: In accordance with ASTM D1480-12 for testing of density or relative density. A certificate of calibration is issued in accordance with the requirements of ISO/IEC 17025.

Specifications

Specifications

  • Pk:
    100 mL
  • Reference temperature:
    20 °C
  • Description:
    Density standard
  • Density:
    0,7374 g/ml

Specifications