Order Entry
Export
Orders LinkContactUsLinkComponent
Wipes for delicate tasks, Kimtech™ Science

Wipes for delicate tasks, Kimtech™ Science

Delicate task wipes, Kimtech™ Science, 7558, 2 ply, 205×200 mm, 15 boxes of 200 wipes
Catalog # 115-2075
Supplier:   KIMBERLY CLARK
Wipes for delicate tasks, Kimtech™ Science
Catalog # 115-2075
Supplier:  KIMBERLY CLARK
Wipes for delicate tasks, Kimtech™ Science
Wipes for delicate tasks, Kimtech™ Science

Delicate task wipes, Kimtech™ Science, 7558, 2 ply, 205×200 mm, 15 boxes of 200 wipes

Price
148,00 €
Log in for availability
1 * 3.000 items
Product Details & Documents

These high quality wipes, made from 100% pure fibre, are suitable for a variety of precision wiping tasks such as the cleaning of delicate glassware, equipment and surfaces. They are also suitable for cleaning liquids and dust, and are equipped with anti-static dispensing to reduce lint and anti-static discharge.

  • Light-duty wipes
  • Low lint absorbency for delicate applications and surfaces
  • Chemically inert and anti-static
  • Supplied in a hygienic, user friendly pop-up box for clean and fast dispensing, even while on the move

Ideal for applications in healthcare, manufacturing, optics and electronics. Suitable for use in ISO Class 4 or higher cleanroom environments.

Colour: White

Specifications
  • Packed:
    15 boxes of 200 wipes
  • Description:
    Delicate task wipes, Kimtech™ Science, 7558, 2 ply
  • L×W:
    205×200 mm
  • Seq:
    02