Orders LinkContactUsLinkComponent
CF®DI Hydrazide
CF®DI Hydrazide
CF®660DI Hydrazide, 661/703 nm, 1 mg
Catalog # 92184.
Supplier:   Biotium
CF®DI Hydrazide
Catalog # 92184.
Supplier:  Biotium
CAS Number:  
CF®DI Hydrazide
CF®DI Hydrazide
CF®660DI Hydrazide, 661/703 nm, 1 mg
CAS Number:  
Price
308,00 €
Log in for availability
1 * 1 mg
Product Details & Documents
CF®DI dye hydrazides react with molecules containing aldehyde or ketone groups to form a hydrazone bond. The dyes are size- and charge-matched and do not significantly alter the molecular weight or pI of the labeled proteins. They are used for detection of protein carbonylation by fluorescence 2D-DIGE.
  • React with carbonyl moieties of aldehyde or ketone groups.
  • Size- and charge-matched, do not significantly alter the molecular weight or pI of the labeled protein.
  • For detection of protein carbonylation by fluorescence 2D-DIGE.
  • Available in several colors for detection of multiple samples on the same gel.
Also see our regular CF® Dye Hydrazides, available in a wide selection of colors.
Specifications
Specifications
  • Pk:
    1 mg
  • Description:
    CF®660DI Hydrazide
  • Ex/Em:
    661/703 nm
Specifications