Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
OBJECTIVE N-ACHROPLAN 10X/0.25 WD6.5 M27 1 * 1 items
Catalog # ZEIS4209409901000
Supplier:
ZEISS
Expand image
OBJECTIVE N-ACHROPLAN 10X/0.25 WD6.5 M27 1 * 1 items
Catalog # ZEIS4209409901000
Supplier:
ZEISS