Organic reference standards, imatinibs, Ph. Eur.
Imatinib impurity H, 1-(pyridin-3-yl)ethan-1-one, Ph. Eur. standard, Reference standard, 130 mg
Catalog # EDQMY0001694
Supplier: EDQM
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:130 mg
- Standards:Reference standard
- Description:Imatinib impurity H, 1-(pyridin-3-yl)ethan-1-one, Ph. Eur. standard
- Cat. No.:EDQMY0001694
- Formula:C₇H₇NO
- Description:Imatinib impurity H, 1-(pyridin-3-yl)ethan-1-one
- CAS No.:350-03-8



