Organic reference standards, imatinibs, Ph. Eur.
Imatinib impurity F, 4-methyl-N3-[4-(pyridin-3-yl)pyrimidin-2-yl]benzene-1,3-diamine, Ph. Eur. standard, Reference standard, 10 mg
Catalog # EDQMY0001693
Supplier: EDQM
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:10 mg
- Standards:Reference standard
- Description:Imatinib impurity F, 4-methyl-N3-[4-(pyridin-3-yl)pyrimidin-2-yl]benzene-1,3-diamine, Ph. Eur. standard
- Cat. No.:EDQMY0001693
- Formula:C₁₆H₁₅N₅
- Description:Imatinib impurity F, 4-methyl-N3-[4-(pyridin-3-yl)pyrimidin-2-yl]benzene-1,3-diamine
- CAS No.:152460-10-1



