Mop frame, aluminium
Mop frame, aluminium, ClinoConnect lock, 400 mm
Catalog # PPSA2009161
Supplier: PFENNIG REINIGUNGSTECHNIK
Jump to:
Product Details & Documents
Aluminium mop frame with reduced weight.
- Mechanics made of stainless steel, plate made of aluminium
- Autoclavable
- Contactless mop drop with MopDrop
Specifications
- Description:Mop frame, aluminium, ClinoConnect lock
- Length:400 mm
- Seq:03