Objectives for CKX53
C plan achromatic objective, 10,5 mm
Catalog # OLYMN4239600
Supplier: OLYMPUS OPTICAL
Jump to:
Product Details & Documents
Specifications
- Magnification:10×/0,17
- Description:C plan achromatic objective
- Working distance:10,5 mm