ContactUsLinkComponent
MidiKros 20cm MPES 750kD 1,0mm 1/2TC X FLL sterile 1 * 1 items
Catalog # SPECT02-E750-10-S
MidiKros 20cm MPES 750kD 1,0mm 1/2TC X FLL sterile 1 * 1 items
Catalog # SPECT02-E750-10-S