Orders LinkContactUsLinkComponent
Milk butyrometer
Butyrometer for milk, 0 - 12% : 0,1
  GERB3157-C-O
Milk butyrometer
  GERB3157-C-O
Milk butyrometer
Milk butyrometer
Butyrometer for milk, 0 - 12% : 0,1
21,30 €
1 * 1 items
Borosilicate glass.
  • With or without stopper
Available with a declaration of conformity per batch and type (in German only).
  • Description:
    Butyrometer for milk, 0 - 12% : 0,1
  • Seq:
    13
Accessories
  •  
    Description
  • 614-2028
    Stopper, rubber, conical, for all measuring method butyrometers
    1 * 100 items - 48,20 €