Orders LinkContactUsLinkComponent
Cold trap, type KF54V
Cold trap, type KF54V
Cold trap, KF 54V-K16-Z-DSS2000, stainless steel, with 200 ml condensate capacity and 1200 ml coolent cap
Catalog # KGWA17111
Supplier:   KGW
Cold trap, type KF54V
Catalog # KGWA17111
Supplier:  KGW
Cold trap, type KF54V
Cold trap, type KF54V
Cold trap, KF 54V-K16-Z-DSS2000, stainless steel, with 200 ml condensate capacity and 1200 ml coolent cap
Price
1 440,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Cold traps are used in conjunction with vacuum pumps to collect condensation produced from humidity or solvents.
  • Easy to clean
  • No stand material for holding the cold trap
  • Protective casing of Dewar flask
Specifications
  • Description:
    Cold trap, KF 54V-K16-Z-DSS2000, stainless steel, with 200 ml condensate capacity and 1200 ml coolent cap
  • Seq:
    03