Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Recombinant, Human. (PLA2, sPLA2-IIA) 1 * 10 µG
Catalog # BMOLP4074-06E2.10
Supplier:
BIOMOL RESEARCH LABORATORIES
Expand image
Recombinant, Human. (PLA2, sPLA2-IIA) 1 * 10 µG
Catalog # BMOLP4074-06E2.10
Supplier:
BIOMOL RESEARCH LABORATORIES