Overalls, PP, ST11
Dark blue, L
Catalog # PRTWST11NARL
Supplier: Portwest
Jump to:
Product Details & Documents
This disposable lightweight overall are made from a breathable non woven PP fabric, enabling the user to keep cool while wearing the garment.
- Generous size design provides exceptional comfort
- Shields against non-hazardous dirt, dust and grime
Weight: 40 g
Specifications
- Clothing Size:L
- Colour:Dark blue
- Seq:08
- Cat. No.:PRTWST11NARL
- Model:ST11
- Garment application:General Purpose|Food
- Weight:40 g/m²