Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CUP OF QUARTZ GLASS , 2000ML , D. 160MM X 140MM HEIGHT , STRAIGHT BOTTOM , STRAIGHT EDGE 1 * 1 items
Catalog # NEUB8450-11-200016
Supplier:
NEUBERT VOLUME GLASSWAERE
Expand image
CUP OF QUARTZ GLASS , 2000ML , D. 160MM X 140MM HEIGHT , STRAIGHT BOTTOM , STRAIGHT EDGE 1 * 1 items
Catalog # NEUB8450-11-200016
Supplier:
NEUBERT VOLUME GLASSWAERE