Cut resistant gloves, A621
L, Black/grey
Catalog # PRTWA621BKRL
Supplier: Portwest
Jump to:
Product Details & Documents
Cut resistance combined with superb grip in oily conditions. ³/₄ nitrile foam dipping absorbs oil keeping hands clean and dry to ensure secure handling.
- ANSI cut level A3
- Materials: HDPE, glass fibre, nitrile foam
- 100% breathable seamless liner
- 13 gauge liner for a perfect fit
- Level D cut resistance with abrasion and tear resistance
Nitrile foam coating for excellent grip in wet and dry conditions.
Certifications: Standards: AS/NZS 2161 .2, AS/NZS 2161 .3, AS/NZS 2161 .4, ANSI/ISEA 105: 2016 CUT Level A4, EN388:2016 +A1:2018 - 4.X.4.3.D., EN 407 X.1.X.X.X.X., EN 420.
Specifications
- Glove size:L
- Colour:Black/grey
- Seq:03
- Cat. No.:PRTWA621BKRL
- Ambidextrous / hand-specific:Hand-specific