Valves for HPLC
Bottom vent valve for HPLC, 1/16" fittings, 1/4 - 28, stainless steel, includes nuts and ferrules
REST26402
: Restek
These valves have low internal volumes and are rated to 15000 psi. They feature a two-piece stem assembly in which the rotating upper shank is coupled through a ball joint to a static lower stem.
- Only fingertight torque is required to make the seal
- Description:Bottom vent valve for HPLC, 1/16" fittings, 1/4 - 28, stainless steel, includes nuts and ferrules
- Seq:03