Orders LinkContactUsLinkComponent
Packed GC columns, 2abc refinery column set
Packed GC columns, 2abc refinery column set
Analytical controls configuration
Catalog # REST88000-810
Supplier:   Restek
Packed GC columns, 2abc refinery column set
Catalog # REST88000-810
Supplier:  Restek
Packed GC columns, 2abc refinery column set
Analytical controls configuration
Price
2 371,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Special columns for unsaturated light hydrocarbons.
  • Faster separations of C1 to C4 hydrocarbons
When used in the proper valving system, it will elute C5+ hydrocarbons ahead of C1 through C4 hydrocarbons.
Specifications
  • Description:
    Analytical controls configuration
  • Seq:
    01