Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MXT-S-BOND Column 2 x 15m, 0.53mm ID, 20um w/ 3.5mm ID without cage 1 * 1 items
Catalog # REST79712-111273
Supplier:
Restek
Expand image
MXT-S-BOND Column 2 x 15m, 0.53mm ID, 20um w/ 3.5mm ID without cage 1 * 1 items
Catalog # REST79712-111273
Supplier:
Restek