Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MXT-50 Cap. Column 60m, 0.53mm ID, 1.50um 3.5 OD without cage 1 * 1 items
Catalog # REST70573-273
Supplier:
Restek
Expand image
MXT-50 Cap. Column 60m, 0.53mm ID, 1.50um 3.5 OD without cage 1 * 1 items
Catalog # REST70573-273
Supplier:
Restek