Orders LinkContactUsLinkComponent
HPLC maintenance kits
HPLC maintenance kits
Ferrules, 1/16"
Catalog # REST26548
Supplier:   Restek
HPLC maintenance kits
Catalog # REST26548
Supplier:  Restek
HPLC maintenance kits
HPLC maintenance kits
Ferrules, 1/16"
Price
73,46 €
Log in for availability
1 * 10 items
Product Details & Documents
These kits are used for start-up and maintenance in all HPLC systems.
PEEK kits contain tubing, fittings, and tools essential for setting up and maintaining your HPLC system: PEEK tubing, connectors, and elbows, PTFE tubing, a tubing cutter and extra blades, a ValvTool wrench, open-end wrenches and more.
Stainless steel kits contain a wide range of tubing, fittings, and tools necessary to set up and maintain your HPLC system: a selection of lengths and IDs of ¹/₁₆" tubing, plus nuts, ferrules, a ValvTool wrench, and a zero-dead-volume union.
Specifications
  • Description:
    Ferrules, 1/16"
  • Seq:
    05