QuEChERS extraction salt packets Q-sep™
Q-sep QuEChERS extraction Q150 packets (AOAC 2007.01 methods) with 6 g magnesium sulphate and 1,5 g sodium acetate
REST26238
: Restek
Jump to:
Q-sep™ QuEChERS provides fast, simple sample preparation for multi residue pesticide analysis.
- Preweighed, ultra-pure sorbents
- Convenient and easy to use
- Salt packets eliminate the need for a second empty tube to transfer salts
Support original unbuffered, AOAC (2007.01) and European (EN 15662) QuEChERS methods.
Specifications
- Description:Q-sep QuEChERS extraction Q150 packets (AOAC 2007.01 methods) with 6 g magnesium sulphate and 1,5 g sodium acetate
- Seq:05