Orders LinkContactUsLinkComponent
QuEChERS extraction salt packets Q-sep™
QuEChERS extraction salt packets Q-sep™
Q-sep QuEChERS extraction Q150 kit (AOAC 2007.01 method) includes 6 g magnesium sulphate, 1,5 g sodium acetate and 50 ml centrifuge tube
Catalog # REST26237
Supplier:   Restek
QuEChERS extraction salt packets Q-sep™
Catalog # REST26237
Supplier:  Restek
QuEChERS extraction salt packets Q-sep™
QuEChERS extraction salt packets Q-sep™
Q-sep QuEChERS extraction Q150 kit (AOAC 2007.01 method) includes 6 g magnesium sulphate, 1,5 g sodium acetate and 50 ml centrifuge tube
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents
Q-sep™ QuEChERS provides fast, simple sample preparation for multi residue pesticide analysis.
  • Preweighed, ultra-pure sorbents
  • Convenient and easy to use
  • Salt packets eliminate the need for a second empty tube to transfer salts
Support original unbuffered, AOAC (2007.01) and European (EN 15662) QuEChERS methods.
Specifications
  • Description:
    Q-sep QuEChERS extraction Q150 kit (AOAC 2007.01 method) includes 6 g magnesium sulphate, 1,5 g sodium acetate and 50 ml centrifuge tube