Orders LinkContactUsLinkComponent
Acylation derivatisation reagents
Acylation derivatisation reagents
PFPOH (pentafluoropropanol), 25 g, 1 Ampoul
Catalog # REST35625
Supplier:   Restek
Acylation derivatisation reagents
Catalog # REST35625
Supplier:  Restek
CAS Number:  
Acylation derivatisation reagents
PFPOH (pentafluoropropanol), 25 g, 1 Ampoul
CAS Number:  
Price
288,89 €
Log in for availability
1 * 1 Ampoul
Product Details & Documents
Acylation reagents offer the same advantage available from silylation reagents: creating less polar, more volatile derivatives. In comparison to silylating reagents, the acylating reagents can more readily target highly polar multifunctional compounds, such as carbohydrates and amino acids. In addition, acylating reagents offer the distinct advantage of introducing electron-capturing groups, thus enhancing detectability during analysis.
  • Most commonly used for electron capture detection
  • React with alcohols, amines, and phenols
  • Frequently used for drugs of abuse confirmation
Specifications
Specifications
  • Pk:
    1 Ampoul
  • Capacity:
    25 g
  • Description:
    PFPOH (pentafluoropropanol)
Specifications