Chloroform ≥98% stabilised, TECHNICAL
Metal drum, 200 L
Supplier: VWR Chemicals
Chloroform ≥98% stabilised, TECHNICAL
Catalog # 22720.550
Supplier: VWR Chemicals
CAS Number: 67-66-3
Jump to:
Product Details & Documents
Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
- CAS Number:67-66-3
- Formula:CHCl₃
- MDL Number:MFCD00000826
- Molecular Weight:119.38 g/mol
- Merck Index:13,02160
- Boiling Pt:61.7 °C (1013 hPa)
- Melting Pt:–63 °C
- Density:1.4832 g/cm³ (20 °C)
- EINECS:200-663-8
- ADR:6.1,III
- :Stabilised with ethanol 0.6 %
- Storage Temperature:Ambient
- UN:1888
- REACH:01-2119486657-20
Specifications
Specification Test Results
- Assay:Min. 98 %
- Density (20/4):1.473 - 1.489
- Ethanol:0.1 - 1.0 %
Specifications
- Pack type:Metal drum
- Pk:200 L



