Order Entry
Export
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Sep-Pak® Vac

Solid phase extraction cartridges, Sep-Pak® Vac

Cartridge, Silica, 55 - 105 µm, 1,2 ml, 500 mg
Catalog # WATEWAT020810
Supplier:   WATERS
Solid phase extraction cartridges, Sep-Pak® Vac
Catalog # WATEWAT020810
Supplier:  WATERS
Solid phase extraction cartridges, Sep-Pak® Vac

Cartridge, Silica, 55 - 105 µm, 1,2 ml, 500 mg

Price
328,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These cartridges deliver the option of minimum sample preparation. Consists of moulded, high purity PP syringe-barrel bodies with two PE frits to contain the packing material.

  • Standard male Luer permits use with multi-position vacuum manifolds
  • Colour coded printing on syringe barrels for identification
Specifications
  • Column Format:
    Cartridge
  • Particle Size:
    55 - 105 µm
  • Capacity:
    1,2 ml
  • Seq:
    09
  • Bed weight:
    500 mg
  • Phase:
    Silica