Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ACE 3 C4-300 NANO COLUMN 150 x 0.075mm 1 * 1 items
Catalog # ACE2131500075
Supplier:
Avantor
Expand image
ACE 3 C4-300 NANO COLUMN 150 x 0.075mm 1 * 1 items
Catalog # ACE2131500075
Supplier:
Avantor