Digital block heater, with touch screen
Block heater, touch screen, 4, 600 W, RT* +5...130 °C, ≤±1 °C, 535×200×100 mm, 5,1 kg
Catalog # 460-0311
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Designed to provide precise temperature stability and uniformity. The units have an easy to use touch screen control which displays time and temperature and are durable with a powder-coated steel body. Ideal for sample preparation, enzyme preservation, enzyme-substrate reactions, DNA amplification, electrophoresis gel degeneration and serum coagulation. Choice of models to hold 1, 2 and 4 interchangeable modular blocks to accommodate a variety of vessels and applications. Interchangeable aluminum alloy sample blocks designed to provide versatility, easy cleaning and disinfecting.
- Built-in temperature sensing probe designed for outstanding temperature accuracy and control
- Temperature deviation adjustment
- Precise temperature control with PID circuit
- Temperature calibration enables user to offset the temperature to desired value
- Timer enables the user to accurately monitor the heating time
- 10 user setting programmes, each with up to 5 sequential steps of different temperatures and duration times
: Supplied with block extraction tool, but without blocks, these must be ordered separately.
: * RT = Ambient
Note: Temp. stability value shown above is at 37 °C.
Specifications
- Heating capacity:600 W
- No. of blocks:4
- Temperature Range:RT* +5...130 °C
- Weight:5,1 kg
- Seq:03
- W×D×H:535×200×100 mm
- Temp. stability:≤±1 °C
- Type:Block heater, touch screen
Accessories
Product Information
- Catalog #AvailabilityPrice
- PT1000 temperature probe
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium blockAluminium block460-029825 mm + 12-13 mm + 6 mm Ø tubes3+12+6Log in for availability1 * 1 items - 172,00 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Mixed aluminium blockMixed aluminium block460-02966 mm + 10 mm Ø tubes32+21Log in for availability1 * 1 items - 126,00 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForW×D×HAvailabilityPrice
- Aluminium blockAluminium block460-029596-well microplate124×76×39 mmLog in for availability1 * 1 items - 88,90 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium blockAluminium block460-03051×96 well PCR plate, half or full skirted-Log in for availability1 * 1 items - 266,00 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium blockAluminium block460-03041×96-well PCR plate, non skirted-Log in for availability1 * 1 items - 232,00 €
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block
Product Information
- Catalog #ForNo. of HolesAvailabilityPrice
- Aluminium block



