Order Entry
Export
Orders LinkContactUsLinkComponent
 

Super Adjustable™ Super Erecta® stem castor carts

Super Adjustable™ Super Erecta® stem castor cart, A366BC, 4-tier model, with 2-swivel and 2-brake resilient rubber castors, Chrome plated, 457×1524×1753 mm, 40 kg
Catalog # METOA366BC
Supplier:   Metro International
Super Adjustable™ Super Erecta® stem castor carts
Catalog # METOA366BC
Super Adjustable™ Super Erecta® stem castor carts
Super Adjustable™ Super Erecta® stem castor carts

Super Adjustable™ Super Erecta® stem castor cart, A366BC, 4-tier model, with 2-swivel and 2-brake resilient rubber castors, Chrome plated, 457×1524×1753 mm, 40 kg

Price
1 260,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Designed to work in conjunction with the entire Super Erecta® system of shelves and accessories. These Super adjustable wire shelves have a unique corner release system which allows them to be adjusted quickly and easily without the use of tools.

  • Allow for quick and easy adjustability
  • Strong and versatile
  • 5- or 4-tier models
  • Resilient rubber or polyurethane castors with donut bumpers

Shelf has a sleek chrome finish that makes it perfect for dry storage areas, while the open-wire construction minimises dust accumulation and allows a free circulation of air and greater visibility.

Chrome shelves and chrome-plated posts.

Specifications
  • Description:
    Super Adjustable™ Super Erecta® stem castor cart, A366BC, 4-tier model, with 2-swivel and 2-brake resilient rubber castors
  • Material:
    Chrome plated
  • Seq:
    01
  • W×D×H:
    457×1524×1753 mm
  • Weight:
    40 kg