Volumetric flasks, class A, white markings
100 ml, 12/21, ± 0,10 ml
POULUS258/AM/WACG5
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
: Acc. to ISO and ASTM or USP standards
- NS:12/21
- Capacity:100 ml
- Seq:31
- Tolerance:± 0,10 ml



