Volumetric flasks, class A, white markings
50 ml, 12/21, ± 0,06 ml
Catalog # POULUS258/AM/WACF5
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass, amber, with PP NS-stopper.
- Easy to read white markings
- Very good chemical resistance
- Very good temperature resistance
Amber coloured to protect light sensitive substances.
: Acc. to ISO and ASTM or USP standards
Specifications
- NS:12/21
- Capacity:50 ml
- Seq:30
- Tolerance:± 0,06 ml



