Order Entry
Export
Orders LinkContactUsLinkComponent
 

Volumetric flasks, class A, white markings

20 ml, 10/19, ± 0,04 ml
Catalog # POULUS258/AM/WACD5
Supplier:   Poulten & Graf
Volumetric flasks, class A, white markings
Catalog # POULUS258/AM/WACD5
Supplier:  Poulten & Graf
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings

20 ml, 10/19, ± 0,04 ml

Price
137,00 €
Log in for availability
1 * 5 items
Product Details & Documents

Borosilicate glass, amber, with PP NS-stopper.

  • Easy to read white markings
  • Very good chemical resistance
  • Very good temperature resistance

Amber coloured to protect light sensitive substances.

 : Acc. to ISO and ASTM or USP standards

Specifications
  • NS:
    10/19
  • Capacity:
    20 ml
  • Seq:
    28
  • Tolerance:
    ± 0,04 ml