Safety Spectacles, Silium+
Anti-UV (5-3,1), anti-fog, anti-scratch, Anthracite, Smoke PC
Catalog # 111-1627
Supplier: Bollé Safety
Jump to:
Product Details & Documents
Inspired from SILIUM, SILIUM + are comfortable and ultra lightweight PC safety glasses. SILIUM + is an excellent choice, combining style, ergonomics and performances for extended use.
- Adjustable bridge
- Non-slip bridge
- Straight, highly flexible ergonomic temples
- PLATINUM coating
Weight: 21 g
Certifications: EN 166 FT1, EN 170, EN 172 (smoke lens).
Documents
BOLESILPPSI_TDS_EN
Datasheet
Specifications
- Type:Anti-UV (5-3,1), anti-fog, anti-scratch
- Colour:Anthracite
- Seq:02
- Lenses:Smoke PC