Order Entry
Export
Orders LinkContactUsLinkComponent
 

Master mixes, green multiplex, Phusion® U

Phusion® U green multiplex PCR master mix, 100 reactions (50 μl), 1 KIT
Catalog # F-564S
Supplier:   FINNZYMES REAGENTS
Master mixes, green multiplex, Phusion® U
Catalog # F-564S
Supplier:  FINNZYMES REAGENTS
CAS Number:  
Master mixes, green multiplex, Phusion® U
Master mixes, green multiplex, Phusion® U

Phusion® U green multiplex PCR master mix, 100 reactions (50 μl), 1 KIT

CAS Number:  
Price
295,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Phusion® U green multiplex PCR master mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube.

  • Specific and efficient multiplex PCR of over 20 targets up to 2,5 kb
  • Excellent results with templates of suboptimal purity due to inhibitor tolerance
  • Fast cycling and direct loading of PCR products on gels
  • Store at –20 °C

Over 20 primer pairs may be combined into a single reaction for highly specific and efficient multiplexing over a broad range of primer and template concentrations.

It contains a proprietary buffer with balanced concentrations of all PCR components eliminating the need for tedious optimization. The Green format further simplifies the workflow - it includes a density reagent and two electrophoresis tracking dyes for direct loading of PCR products on gels.

Applications: genotyping, pathogen detection, food testing, analysis of genetically modified organisms, amplification of microsatellites.

Similarly to other enzymes from Phusion® family, Phusion® U DNA polymerase incorporates more nucleotides per binding event than any other DNA polymerase. This allows achieving high yields of PCR products with shorter extention times and consequently reduced total protocol times. Significantly enhanced processivity also enables successful multiplex PCR on difficult targets such as GC-rich templates or templates of suboptimal purity. Being highly tolerant of many PCR inhibitors, it can even be used with unpurified samples such as blood or serum.

Specificity of multiplex PCR is increased by affibody-mediated hot start mechanism. Reversibly bound affibody ligand inhibits the polymerase at ambient temperatures preventing amplification of nonspecific products and formation of primer-dimers.

 : Includes: Phusion® U hot start DNA polymerase, 2X reaction buffer, 3 mm MgCl₂, 400 µm of each dATP, dTTP, dCTP and dGTP. The master mix is supplied with nuclease-free water.

 : For research use only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    Phusion® U green multiplex PCR master mix, 100 reactions (50 μl)

Specifications