Order Entry
Export
Orders LinkContactUsLinkComponent
 

GC liners, LinerGOLD™

LinerGOLD split, 6,3 mm, 1,0 mm, 78,5 mm
  HYPE453A1335-UI
GC liners, LinerGOLD™
  HYPE453A1335-UI
GC liners, LinerGOLD™
GC liners, LinerGOLD™

LinerGOLD split, 6,3 mm, 1,0 mm, 78,5 mm

 
135,00 €
   
1 * 5 items
 

LinerGOLD™ GC liners provide exceptional inertness, leading to increased accuracy and precision by preventing loss of sensitive compounds.

  • Lower detection limits for a wide range of active compounds
 
  • Ext.Ø:
    6,3 mm
  • Ø int.:
    1,0 mm
  • Description:
    LinerGOLD split
  • Length:
    78,5 mm
  • Seq:
    11