Accessories
UBS 2 conversion kit for load carriers and upper baskets of G 78 series, without drying connection, PG 85
Catalog # MIEL10131100
Supplier: MIELE
Jump to:
Product Details & Documents
- For lower and upper baskets
- With 7 x 7 mm perforations
- Stainless steel
Specifications
- Description:UBS 2 conversion kit for load carriers and upper baskets of G 78 series, without drying connection
- Seq:44
- For:PG 85