0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Buffer Solution pH 9.180 1 * 1 L
Buffer Solution pH 9.180 1 * 1 L
APPC277123.1211
:
PanReac AppliChem
Buffer Solution pH 9.180 1 * 1 L
APPC277123.1211
:
PanReac AppliChem
Buffer Solution pH 9.180 1 * 1 L
66,30 €
1 * 1 L
-
+