0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Buffer Solution pH 2.00 (20°C) 1 * 1.000 mL
Buffer Solution pH 2.00 (20°C) 1 * 1.000 mL
APPC272581.1211
:
PanReac AppliChem
Buffer Solution pH 2.00 (20°C) 1 * 1.000 mL
APPC272581.1211
:
PanReac AppliChem
Buffer Solution pH 2.00 (20°C) 1 * 1.000 mL
43,80 €
1 * 1.000 mL
-
+