Oleic acid
1.000 mL
Supplier: PanReac AppliChem
Jump to:
Product Details & Documents
Synonyms: (Z)-9-Octadecenoic acid.
Synonyms: (9Z)-9-Octadecenoic acid
- CAS Number:112-80-1
- Formula:C₁₈H₃₄O₂
- MDL Number:MFCD00064242
- Molecular Weight:282.47 g/mol
- Boiling Pt:286 °C
- Melting Pt:4 °C
- Density:0.890 kg/l
Documents
Specifications
Specification Test Results
- Assay (G.C.):65,0 - 88,0%
- Appearance:Passes test
- Residue on ignition (as SO₄):0,1%
- Acidity value:195 - 204
- Iodine value:89 - 105
- Peroxide value:10,0
- Residual solvents (Ph. Eur.):Passes test
- Myristic acid:5,0%
- Palmitic acid:16,0%
- Palmitoleic acid:8,0%
- Stearic acid:6,0%
- Linoleic acid:18,0%
- Linolenic acid:4,0%
- Fatty acids with chain length greater than C18:4,0%
- Margaric acid:4,0%
- Identity:IR passes test
- Cadmium (Cd) - Class 1 impurities (ICH Q3D):0,5 ppm
- Lead (Pb) - Class 1 impurities (ICH Q3D):2 ppm
- Arsenic (As) - Class 1 impurities (ICH Q3D):1,5 ppm
- Mercury (Hg) - Class 1 impurities (ICH Q3D):1,5 ppm
- Cobalt (Co) - Class 2A impurities (ICH Q3D):5 ppm
- Vanadium (V) - Class 2A impurities (ICH Q3D):10 ppm
- Nickel (Ni) - Class 2A impurities (ICH Q3D):20 ppm
- Thallium (Tl) - Class 2B impurities (ICH Q3D):5 ppm
- Gold (Au) - Class 2B impurities (ICH Q3D):10 ppm
- Palladium (Pd) - Class 2B impurities (ICH Q3D):10 ppm
- Iridium (Ir) - Class 2B impurities (ICH Q3D):10 ppm
- Osmium (Os) - Class 2B impurities (ICH Q3D):10 ppm
- Rhodium (Rh) - Class 2B impurities (ICH Q3D):10 ppm
- Ruthenium (Ru) - Class 2B impurities (ICH Q3D):10 ppm
- Selenium (Se) - Class 2B impurities (ICH Q3D):15 ppm
- Silver (Ag) - Class 2B impurities (ICH Q3D):15 ppm
- Platinum (Pt) - Class 2B impurities (ICH Q3D):10 ppm
- Lithium (Li) - Class 3 impurities (ICH Q3D):55 ppm
- Antimony (Sb) - Class 3 impurities (ICH Q3D):120 ppm
- Barium (Ba) - Class 3 impurities (ICH Q3D):140 ppm
- Molybdenum (Mo) - Class 3 impurities (ICH Q3D):25 ppm
- Copper (Cu) - Class 3 impurities (ICH Q3D):250 ppm
- Tin (Sn) - Class 3 impurities (ICH Q3D):600 ppm
- Chromium (Cr) - Class 3 impurities (ICH Q3D):25 ppm
Specifications
- Pk:1.000 mL



