Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium Hydroxide solution 50 % pure 1 * 5 L

Potassium Hydroxide solution 50 % pure 1 * 5 L

Catalog # APPC142403.1214
Supplier:   PanReac AppliChem
Potassium Hydroxide solution 50 % pure 1 * 5 L
Catalog # APPC142403.1214
Supplier:  PanReac AppliChem
CAS Number:  
Potassium Hydroxide solution 50 % pure 1 * 5 L
CAS Number:  
Price
137,00 €
Log in for availability
1 * 5 L
Product Details & Documents