Order Entry
Export
Orders LinkContactUsLinkComponent
Flasks, pear shaped, with two necks

Flasks, pear shaped, with two necks

250 ml, 24/29 NS, 24/29 NS, 140 mm
  WITG0.663.250.E
Flasks, pear shaped, with two necks
  WITG0.663.250.E
Flasks, pear shaped, with two necks
Flasks, pear shaped, with two necks

250 ml, 24/29 NS, 24/29 NS, 140 mm

 
18,70 €
   
1 * 1 items
 

Borosilicate glass 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

The conical geometry makes them ideal for small-scale reactions.

 
  • Capacity:
    250 ml
  • Height:
    140 mm
  • Seq:
    04
  • Side necks:
    24/29 NS
  • Centre neck:
    24/29 NS