ContactUsLinkComponent
Pipette, 8channel, volume 20-200µl, type EP8 200, electronic, division 1µl. 1 * 1 items
Catalog # WITG5.402.420.S
Supplier:  witeg Labortechnik
Pipette, 8channel, volume 20-200µl, type EP8 200, electronic, division 1µl. 1 * 1 items
Catalog # WITG5.402.420.S
Supplier:  witeg Labortechnik