Order Entry
Export
Orders LinkContactUsLinkComponent
 

7-Ethyl-10-hydroxycamptothecin ≥98%

50 mg
Catalog # CAYM15632-50
Toxic
Health
Supplier:   Cayman Chemical
7-Ethyl-10-hydroxycamptothecin ≥98%
Catalog # CAYM15632-50
Supplier:  Cayman Chemical
CAS Number:  86639-52-3
7-Ethyl-10-hydroxycamptothecin ≥98%

50 mg

CAS Number:  86639-52-3
Price
501,00 €
Log in for availability
1 * 50 mg
Product Details & Documents

SN-38 is the active metabolite of irinotecan, a derivative of the alkaloid camptothecin and acts as a potent inhibitor of DNA topoisomerase I. SN-38 is a more potent topopisomerase I inhibitor and more cytotoxic to HT-29 colon cancer cells (IC50 = 8.8 nM) compared to irinotecan (IC50 >100 nM).

  • SN-38 is in a form of crystalline solid
  • λmax at 224, 267, 331, 368, 383 nm
Synonyms:  10-hydroxy-7-ethylcamptothecin, (S)-4,11-Diethyl-4,9-dihydroxy-1H-pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 7-Ethyl-10-hydroxy-20(S)-Camptothecin, SN-38, NK 012
  • CAS Number:86639-52-3
  • Formula:C₂₂H₂₀N₂O₅
  • MDL Number:MFCD00871873
  • Molecular Weight:392.40 g/mol
  • Storage Temperature:Freezer
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pk:
    50 mg

Specifications