Orders LinkContactUsLinkComponent
Accessories for microcentrifuges, mySPIN™
Accessories for microcentrifuges, mySPIN™
12×1,5/2,0 ml fixed angle tube rotor, reinforced glass fiber, max. RCF 9800×g, max. speed 12500 min⁻¹ for microcentrifuge mySPIN™ 12
Catalog # HERA75004083
Supplier:   Thermo Scientific
Accessories for microcentrifuges, mySPIN™
Catalog # HERA75004083
Supplier:  Thermo Scientific
Accessories for microcentrifuges, mySPIN™
Accessories for microcentrifuges, mySPIN™
12×1,5/2,0 ml fixed angle tube rotor, reinforced glass fiber, max. RCF 9800×g, max. speed 12500 min⁻¹ for microcentrifuge mySPIN™ 12
Price
83,20 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    12×1,5/2,0 ml fixed angle tube rotor, reinforced glass fiber, max. RCF 9800×g, max. speed 12500 min⁻¹ for microcentrifuge mySPIN™ 12
  • Seq:
    03